Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Janusin Mouse, Unlabeled, Clone: 9, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610629

Catalog No. BDB610629

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Store undiluted at -20°C.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

Janusin
9
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Monoclonal
Immunofluorescence, Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit