Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JBP1/Skb1Hs Mouse, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611538

 View more versions of this product

Catalog No. BDB611538


Add to cart

Description

Description

p21[Cdc42/Rac]-activated kinases (PAKs) bind and are activated by the small GTPases, Cdc42 and Rac. In yeast, the interaction of the PAK, Shk1, with Skb1 (Shk1 kinase binding protein 1) is important in the regulation of cell morphology and mitosis. Skb1Hs, the human homolog of Skb1, restores growth rates in yeast cells deficient for Skb1. Skb1Hs has also been identified as a 72kDa pICln-binding protein (IBP72). pICln is thought to be a cytosolic regulator of a swelling-induced chloride channel and interactions with Skb1Hs suggest a link with PAKs, which are involved in cytoskeletal rearrangement. An additional line of study has identified Skb1Hs as a Jak binding protein (JBP1) via its interactions with Jak1-3 and Tyk2. This study also identified Skb1Hs as a methyltransferase that interacts with multiple proteins which possibly serve as physiological targets. Skb1Hs is expressed in a wide range of tissues and is thought to form homodimeric or multimeric complexes. Thus, Skb1Hs (IBP72, JBP1) is a multifunctional protein that may participate in such diverse processes as transcriptional and translational regulation, cytoskeletal rearrangement, and any of a number of signaling pathways.

Host Species: Mouse
Clone: 32
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Skb1Hs aa. 423-624
Formula Weight [Chemical]: 72kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

JBP1/Skb1Hs
32
Unconjugated
Affinity Purified
Human Skb1Hs aa. 423-624
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.