Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JIK Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513735

Catalog No. PIPA513735

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken, mouse, rat and Xenopus based on sequence homology.

TAOK3 is negatively regulated by epidermal growth factor (EGF) and Inhibits the basal activity of Jun kinase. When TAOK3 is overexpressed, it may activate ERK1/ERK2 and JNK/SAPK.
Specifications

Specifications

JIK
Polyclonal
Unconjugated
TAOK3
Liquid
TAOK3
KLH conjugated synthetic peptide between 671-700 amino acids from the C-terminal region of human JIK (TAOK3)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide
Q9H2K8
EC 2.7.11.1; Thousand and one amino acid protein 3; Jun kinase-inhibitory kinase; JNK/SAPK-inhibitory kinase; Dendritic cell-derived protein kinase; Cutaneous T-cell lymphoma tumor antigen HD-CL-09; CTCL tumor antigen HD-CL-09; Kinase from chicken homolog A; hKFC-A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51347
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit