Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-JIL-1, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals 49950002
Description
JIL-1 Polyclonal antibody specifically detects JIL-1 in Drosophila samples. It is validated for Western Blot, Chromatin Immunoprecipitation.Specifications
JIL-1 | |
1 mg/mL | |
Western Blot 1:100-1:2000, Chromatin Immunoprecipitation 1:10-1:500 | |
Affinity Purified | |
In vivo generated recombinant protein fragment. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Associates with chromosomes throughout the cell cycle. Localizes to interband regions along the polytene chromosomes and is enriched almost two-fold on the male X chromosome compared to the autosome. |
ChIP assay, Western Blot | |
Unconjugated | |
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative | |
Rabbit | |
IgG | |
0.05 mg | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Drosophila |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only