Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JIP-1 Mouse, Unlabeled, Clone: 50, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611891

 View more versions of this product

Catalog No. BDB611891


Add to cart

Description

Description

The JNK group of MAPKs is activated by a variety of inflammatory cytokines and environmental stressors. Activated JNK phosphorylates many cellular proteins, such as components of the AP-1 transcription factor complex (c-Jun and ATF-2). JNK-interacting proteins (JIPs) bind JNK, MKK7, MLKs, p190RhoGEF, and the Ste20-related protein kinase HPK1. In mouse, alternative splicing of JIP produces multiple splice variants, JIP-1, JIP-1b, JIP-2a, JIP-2b, and JIP-3. The structure of full length JIP consists of two N-terminal acidic regions, a JNK binding domain (JBD), two proline rich regions (PR), and both an SH3 and a phosphotyrosine-binding domain in the C-terminal region. JIP-1 localizes to the tips of neurites in differentiated PC12 cells, and may interact with JNK, MKK7, and MLK to facilitate formation of the JNK activating complex. IB1 is the rat homologue of JIP-1b, a JIP-1 variant that includes a 47 amino acid insert in the C-terminal region. IB1 is found in the nucleus and cytoplasm, and may function as a transactivator of the GLUT2 gene. Thus, JIP-1 and its related isoforms may have multiple functions that involve specific protein-protein interactions.

Host Species: Mouse
Clone: 50
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse JIP-1 aa. 180-384

Immunofluorescence, Western Blotting

Specifications

Specifications

JIP-1
50
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
JNK Interacting Protein
Affinity Purified
Mouse JIP-1 aa. 180-384
150μg
Cell Biology
Primary
Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.