Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-JIP2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133035

Catalog No. NBP133035


Add to cart

Description

Description

JIP2 Polyclonal antibody specifically detects JIP2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

JIP2
1.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
MAPK8IP2
Recombinant protein encompassing a sequence within the C-terminus region of human JIP2. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
C-Jun-amino-terminal kinase-interacting protein 2, homologous to mouse JIP-1, IB-2, IB2JIP2Mitogen-activated protein kinase 8-interacting protein 2, islet-brain 2, Islet-brain-2, JIP-2, JNK MAP kinase scaffold protein 2, JNK MAP kinase scaffold protein JIP2, JNK-interacting protein 2, mitogen-activated protein kinase 8 interacting protein 2, PRKM8 interacting protein-like, PRKM8IPL
Rabbit
IgG
100 ul
Apoptosis
Primary
23542
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only