Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524611

Catalog No. PIPA524611

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Histone demethylase that specifically demethylates  ft.Lys-9 ft. of histone H3, thereby playing a role in histone code. Does not demethylate histone H3  ft.Lys-4 ft., H3  ft.Lys-27 ft., H3  ft.Lys-36 ft. nor H4  ft.Lys-20 ft.. Only able to demethylate trimethylated H3  ft.Lys-9 ft., with a weaker activity than KDM4A, KDM4C and KDM4D. Demethylation of Lys residue generates formaldehyde and succinate.
Specifications

Specifications

JMJD2B
Polyclonal
PBS with 0.09% sodium azide
O94953, Q91VY5
JMJD2B, jmjC domain-containing histone demethylation protein 3B, jumonji domain containing 2B, jumonji domain-containing protein 2B, lysine-specific demethylase 4B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
193796, 23030
Western Blot
Unconjugated
KDM4B
Liquid
KDM4B
KLH conjugated synthetic peptide between 281-306 amino acids from human JMJD2B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit