Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2C Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523065

Catalog No. PIPA523065


Add to cart

Description

Description

Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.

JMJD2C is a member of the Jumonji domain 2 (JMJD2) family and encodes a protein with one JmjC domain, one JmjN domain, two PHD-type zinc fingers, and two Tudor domains. This nuclear protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form. Chromosomal aberrations and increased transcriptional expression of this gene are associated with esophageal squamous cell carcinoma.
Specifications

Specifications

JMJD2C
Polyclonal
Unconjugated
KDM4C
Liquid
KDM4C
A synthetic peptide made to an internal region of the human JMJD2C protein (within residues 450-600).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ChIP assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% sodium azide
Q9H3R0, Q8VCD7
RP11-169L18.1, GASC1, JHDM3C, JMJD2C, bA146B14.1, GASC-1 protein, JmjC domain-containing histone demethylation protein 3C, gene amplified in squamous cell carcinoma 1 protein, jumonji domain containing 2C, jumonji domain-containing protein 2C, lysine-specific demethylase 4C
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23081, 76804
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit