Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524466

Catalog No. PIPA524466

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene likely encodes a histone lysine demethylase. Studies of a similar protein in mouse indicate a potential role for this protein as a tumor suppressor. Alternatively spliced transcript variants have been described.
Specifications

Specifications

JMJD5
Polyclonal
PBS with 0.09% sodium azide
Q8N371
jmjC domain-containing protein 5, jumonji domain-containing protein 5, lysine-specific demethylase 8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79831
Western Blot
Unconjugated
KDM8
Liquid
KDM8
KLH conjugated synthetic peptide between 36-63 amino acids from the N-terminal region of human JMJD5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit