Learn More
JNK2 Mouse anti-Human, Mouse, Rat, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05986
Catalog No. 05-986-MI
Add to cart
Description
Description
Detect JNK2 using this Anti-JNK2 Antibody validated for use in ELISA and western blotting.
Specifications
Specifications
JNK2
Unconjugated
Affinity Purified
Murine
IgG
100μg
Primary
Human, Murine, Rat
ELISA, Western Blot
NM_002752
MAPK9, JNK2B, p54aSAPK, PRKM9, JNK2A, JNK-55, JNK2, JNK2BETA, JNK2ALPHA
Recombinant human JNK2 protein
Immunoaffinity purified
−20°C for 24 months.
Monoclonal