Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-JNK2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB6001297SS

 View more versions of this product

Catalog No. NB6001297SS


Add to cart

Description

Description

JNK2 Polyclonal antibody specifically detects JNK2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

JNK2
0.5 mg/mL
Western Blot 1-3 ug/ml
P45984
c-Jun kinase 2, c-Jun N-terminal kinase 2, EC 2.7.11, EC 2.7.11.24, JNK2A, JNK2BETA, JNK2JNK2ALPHA, JNK-55, Jun kinase, MAP kinase 9, MAPK 9, mitogen-activated protein kinase 9, p54a, p54aSAPK, PRKM9JNK2B, SAPK, Stress-activated protein kinase JNK2
Rabbit
IgG
0.025 mg
Cancer, Hypoxia, Immunology, Innate Immunity, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary
5601
Human
Western Blot
Unconjugated
PBS containing 0.2% gelatin with 0.05% Sodium Azide
Purified
MAPK9
A synthetic peptide corresponding to amino acids 373-389 (KDQPSDAAVSSNATPSQ) of human JNK2 was used as immunogen; GenBank no. gi 21237736 ref NP_002743.3 . This amino sequence is 100% homologous in cow and dog, and 94% in mouse and rat.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Cross-reacts with Human. Not yet tested in other species.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only