Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JTV-1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512487

Catalog No. PIPA512487

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

JTV1 is probable core protein of the multisynthetase complex that serves as a template for the assembly of the supramolecular structure. This protein Mediates ubiquitination of FUBP1 and its degradation by the proteasome.
Specifications

Specifications

JTV-1
Polyclonal
PBS with 0.09% sodium azide
Q13155
Multisynthetase complex auxiliary component p38; Protein JTV-1; PRO0992
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7965
Western Blot
Unconjugated
AIMP2
Liquid
AIMP2
KLH conjugated synthetic peptide between 156-184 amino acids from the central region of human JTV1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit