Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K+ Channel α Mouse, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611248

 View more versions of this product

Catalog No. BDB611248


Add to cart

Description

Description

Cellular excitability is modulated by the precise function of voltage-sensitive ion channels. Large conductance potassium channels (Maxi-K) are unique in that they are sensitive to transmembrane potential and intracellular Ca2+; concentrations. These channels, important for neuronal firing and vascular tone, share many features with voltage-dependent Na+, Ca2+;, and K+ channels. Among these are the S4 region, a motif with a repeated triple sequence of one positively charged amino acid and two hydrophobic amino acids. This region is thought to be the voltage sensor. Maxi-K is subject to complex metabolic control control that also involves G proteins and phosphorylation/dephosphorylation reactions. This type of K+ channel is composed of two subunits, the pore-forming α subunit (hslo) and the regulatory β subunit. The α subunit is subject to direct phosphorylation by cyclic GMP-dependent protein kinase (PKG) and dephosphorylation by protein phosphatase 2A. The fact that the α subunit of Maxi-K channels is a substrate of PKG supports the idea that these channels perform an important function in the cellular response to potent vasodilators, such as nitrocompounds and atrial natiuretic peptide.

Host Species: Mouse
Clone: 32
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Human K+ Channel α Subunit aa. 995-1113
Formula Weight [Chemical]: 125kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

K+ Channel α
32
Unconjugated
Affinity Purified
Human K+ Channel α Subunit aa. 995-1113
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.