Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTI1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524898

Catalog No. PIPA524898

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TTI1
Polyclonal
PBS with 0.09% sodium azide
O43156
KIAA0406, smg-10, TEL2-interacting protein 1 homolog, Tel two interacting protein 1, Tel2 interacting protein 1 homolog, smg-10 homolog, nonsense mediated mRNA decay factor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9675
Western Blot
Unconjugated
TTI1
Liquid
TTI1
KLH conjugated synthetic peptide between 41-69 amino acids from the N-terminal region of human K0406
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit