Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA1609 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524016

Catalog No. PIPA524016


Add to cart

Specifications

Specifications

KIAA1609
Polyclonal
PBS with 0.09% sodium azide
Q6P9B6, Q8K0P3
TLD domain-containing protein KIAA1609
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57707, 74347
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TLDC1
Liquid
TLDC1
KLH conjugated synthetic peptide between 426-456 amino acids from the C-terminal region of human KIAA1609
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit