Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT78 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526855

Catalog No. PIPA526855

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is a member of the type II keratin gene family and encodes a protein with an intermediate filament domain. Keratins are the major structural proteins in epithelial cells, forming a cytoplasmic network of 10 to 12 nm wide intermediate filaments and creating a scaffold that gives cells the ability to withstand mechanical and non-mechanical stresses. The genes of the type II keratin family are located as a gene cluster at 12p13.13. Four pseudogenes of this gene family have been identified.
Specifications

Specifications

KRT78
Polyclonal
PBS with 0.09% sodium azide
Q8N1N4
K5B, Kb40, CK-78, K78, cytokeratin-78, keratin 5b, keratin, type II cytoskeletal 78, keratin-5b, keratin-78, type-II keratin Kb40
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
196374
Flow Cytometry, Western Blot
Unconjugated
KRT78
Liquid
KRT78
KLH conjugated synthetic peptide between 176-203 amino acids from the central region of human K2C78
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit