Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kalinin B1 Mouse, Unlabeled, Clone: 17, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610423

Catalog No. BDB610423


Add to cart

Description

Description

Kalinin is an epithelial cell laminin. It is a structural component of the dermal-epidermal junction of skin and basement membrane. The full molecule is composed of a 200 kDa A subunit, a 155 kDa B2 subunit, and a 140 kDa B1 subunit. Another B1k subunit with a predicted molecular weight of 126.9 kDa is encoded by a 3.9kb gene. The apparent molecular weight of Kalinin B1k is 140 kDa due to glycosylation. Its structure is divided into a long arm and a short arm domain that bind and crosslink with the other kalinin subunits.

Specifications

Specifications

Kalinin B1
17
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit