Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 12 Mouse anti-Human, Clone: 364932, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524110

Catalog No. PIMA524110


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Alternate splicing of this gene results in three transcript variants encoding different isoforms.
Specifications

Specifications

Kallikrein 12
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9UKR0
Kallikrein-like protein 5, KLK-L5,
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
43849
Immunoprecipitation, Neutralization, Western Blot
364932
Unconjugated
KLK12
Lyophilized
KLK12
Mouse myeloma cell line NS0-derived recombit human Kallikrein 12 Ala18-Asn248
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit