Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14239

Catalog No. PA14239


Add to cart

Description

Description

PA1-4239 detects Kallikrein-13 from human samples. PA1-4239 has been successfully used in Western blot procedures. PA1-4239 immunizing peptide corresponds to amino acid residues 262-277 from human Kallikrein-13.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. The novel KLK gene, KLK-L4 (also known as KLK-13) is expressed in a variety of tissues including prostate, salivary gland, breast, and testis. Its expression is regulated by steroid hormones in breast cancer cell lines may be involved in the pathogenesis and/or progression of breast cancer.
Specifications

Specifications

Kallikrein 13
Polyclonal
Unconjugated
KLK13
Liquid
KLK13
Synthetic Peptide: I(262) R K Y E T Q Q Q K W L K G P Q(277)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9UKR3
KLK-13, KLK-L4, Kallikrein-like protein 4
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
26085
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit