Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 15 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14212

Catalog No. PA14212


Add to cart

Description

Description

PA1-4212 detects the Kallikrein-15 protein in human samples. PA1-4212 has been successfully used in Western blot procedures. The PA1-4212 immunogen is a synthetic peptide corresponding to residues D(177) T S C D K S Y PG R L(188) of human Kallikrein-15.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
Specifications

Specifications

Kallikrein 15
Polyclonal
Unconjugated
KLK15
Liquid
KLK15
Synthetic peptide corresponding to residues D(177) T S C D K S Y PG R L(188) of human Kallikrein-15.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9H2R5
KLK-15, ACO protease
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55554
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit