Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513342

Catalog No. PIPA513342

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Kallikreins are a group of serine proteases manifesting diverse physiological functions. The glandular kallikreins are a distinct group of serine proteases with an ability to release vasoactive peptides from kininogen in vitro, although the kininogenase activity of different kallikreins is highly variable. Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin. Many kallikrein genes are differentially expressed in various maligcies. Human glandular kallikrein (hK2; encoded by the KLK2 gene) is an emerging tumor marker for prostate cancer.
Specifications

Specifications

Kallikrein 2
Polyclonal
PBS with 0.09% sodium azide
P20151
Kallikrein 2, Tissue kallikrein 2, Glandular kallikrein-1, hGK-1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3817
Western Blot
Unconjugated
KLK2
Liquid
KLK2
KLH conjugated synthetic peptide between 166-197 amino acids from the C-terminal region of human Kallikrein 2 (KLK2)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit