Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14235

Catalog No. PA14235


Add to cart

Description

Description

PA1-4235 detects Kallikrein-3 from human samples. PA1-4235 has been successfully used in Western blot procedures. PA1-4235 immunizing peptide corresponds to amino acid residues 247-261 from human Kallikrein-3.

PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and is abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predomitly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.
Specifications

Specifications

PSA
Polyclonal
Unconjugated
KLK3
Liquid
KLK3
Synthetic Peptide: V(247) H Y R K W I K D T I V A N P(261)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P07288
KLK-3, Prostate-specific antigen, PSA, Kallikrein-3, Semenogelase, Gamma-seminoprotein, Seminin, P-30 antigen
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
354
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit