Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14234

Catalog No. PA14234


Add to cart

Description

Description

PA1-4234 detects Kallikrein-4 from human samples. PA1-4234 has been successfully used in Western blot procedures. PA1-4234 immunizing peptide corresponds to amino acid residues 242-254 from human Kallikrein-4.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel Kallikrein gene Prostase/KLK-L1 (also known as KLK4) is expressed and is up-regulated by androgens and progestins. KLK-L1 may be involved in the pathogenesis and/or progression of prostate, breast, and possibly other maligcies.
Specifications

Specifications

Kallikrein 4
Polyclonal
Unconjugated
KLK4
Liquid
KLK4
Synthetic Peptide: K(242) F T E W I E K T V Q A S(254)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9Y5K2
KLK-L1, KLK-4, Prostase, Kallikrein-like protein 1, Enamel matrix serine proteinase 1
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
9622
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit