Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513344

Catalog No. PIPA513344

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. In some tissues KLK4 expression is hormonally regulated. The expression pattern of a similar mouse protein in murine developing teeth supports a role for the protein in the degradation of enamel proteins.
Specifications

Specifications

Kallikrein 4
Polyclonal
PBS with 0.09% sodium azide
Q9Y5K2
Prostate; Kallikrein-like protein 1; KLK-L1; Enamel matrix serine proteinase 1;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56640, 9622
Western Blot
Unconjugated
KLK4
Liquid
KLK4
KLH conjugated synthetic peptide between 85-115 amino acids from the central region of human Kallikrein 4 (KLK4)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit