Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14238

Catalog No. PA14238


Add to cart

Description

Description

PA1-4238 detects Kallikrein-5 from human samples. PA1-4238 has been successfully used in Western blot procedures. PA1-4238 immunizing peptide corresponds to amino acid residues 280-292 from human Kallikrein-5.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. The novel KLK gene, KLK-L2 (also known as KLK5) is expressed in breast, brain, and testis and is up-regulated by estrogens and progestins.
Specifications

Specifications

Kallikrein 5
Polyclonal
Unconjugated
KLK5
Liquid
KLK5
Synthetic Peptide: K(280) F T K W I Q E T I Q A N S(293)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9Y337
KLK-5, KLK-L2, Stratum corneum tryptic enzyme, Kallikrein-like protein 2
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
25818
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit