Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14236

Catalog No. PA14236


Add to cart

Description

Description

PA1-4236 detects Kallikrein-9 from human samples. PA1-4236 has been successfully used in Western blot procedures. PA1-4236 immunizing peptide corresponds to amino acid residues 239-250 from human Kallikrein-9.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. (KLK-L3; HGMW-approved symbol KLK9) is a novel kallikrein gene with utility for diagnosis, monitoring and therapeutics of various cancers including those of the breast, prostate and testis.
Specifications

Specifications

Kallikrein 9
Polyclonal
Unconjugated
KLK9
Liquid
KLK9
Synthetic Peptide: H(239) Y L D W I Q E I M E N(250)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9UKQ9
KLK-9, KLK-L3, Kallikrein-like protein 3
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
284366
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit