Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KALRN, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128479

Catalog No. NBP128479


Add to cart

Description

Description

KALRN Polyclonal antibody specifically detects KALRN in Human, Mouse, Rat, Bovine samples. It is validated for Peptide ELISA.
Specifications

Specifications

KALRN
0.5 mg/mL
Peptide ELISA Detection limit 1:64000
NP_001019831.2
ARHGEF24, DUETFLJ16443, DUO, EC 2.7.11.1, FLJ12332, FLJ18623, HAPIP, huntingtin-associated protein interacting protein (duo), Huntingtin-associated protein-interacting protein, Kalirin, kalirin, RhoGEF kinase, Protein Duo, serine/threonine kinase with Dbl- and pleckstrin homology domains, Serine/threonine-protein kinase with Dbl- and pleckstrin homology domain, TRADFLJ18196
Goat
IgG
0.1 mg
Neuroscience
Primary
8997
Bovine, Human, Mouse, Rat
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
KALRN
Peptide with sequence DRGSFRNDGLKAS-C corresponding to N-Terminus according to NP_003938.1, NP_008995.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize reported isoforms NP_001019831.2 and NP_003938.1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only