Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAP Mouse, Unlabeled, Clone: 39, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610334

Catalog No. BDB610334


Add to cart

Description

Description

Proteins that associate with cyclin-dependent kinases may function in cell cycle regulation. Therefore, a two-hybrid screen system was used to isolate cDNA which encode proteins that can interact with Cdk2. One of these was the dual-specificity protein phosphatase known as KAP (Cd k a ssociated p hosphatase). KAP contains the HCXXXXGR motif characteristic of protein tyrosine phosphatases. It utilizes substrates that contain either phosphotyrosine or phosphoserine residues. In addition to binding Cdk2, KAP also associates with Cdc2. KAP does not require a cyclin subunit for binding to Cdk2 and Cdc2. The association of KAP with these two Cdks suggests that KAP may play a role in cell cycle control, possibly by regulating the phosphorylation status of a cdk or of a Cdk-associated protein.

Specifications

Specifications

KAP
39
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Chicken, Frog, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit