Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KAP1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232071

Catalog No. NBP232071


Add to cart

Description

Description

KAP1 Polyclonal antibody specifically detects KAP1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

KAP1
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
NP_035718.2
E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28
Rabbit
IgG
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
TRIM28
The immunogen this antibody was made to, maps to a region between residue 175 to 225 of human Tripartite Motif-Containing 28 using the numbering given in entry NP_035718.2 (GeneID 21849).
Immunogen affinity purified
RUO
Primary
10155
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only