Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAP3A Mouse, Unlabeled, Clone: 14, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610637

Catalog No. BDB610637


Add to cart

Description

Description

KAP3 (kinesin superfamily-associated protein 3) was identified as an accessory protein for the microtubule translocator kinesin and, independently, as SMAP (Smg GDS-associated protein). Alternative splicing of KAP3 mRNA results in proteins of 793 amino acids (KAP3A) and 772 amino acids (KAP3B). KAP3 protein reportedly is highly expressed in brain and testis and displays an apparent molecular weight of 100kDa. Analysis of its primary structure reveals that KAP3A contains nine repeats of the Arm domain (40 amino acids). This domain is characteristic of the Armadillo family of proteins that includes armadillo, β-catenin, plakoglobin, importin, APC, and pp120. In addition, KAP3/SMAP is a substrate for v-src kinase. Phosphorylation of KAP3/SMAP decreases its assocation with Smg GDS.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

KAP3A
14
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Kinesin superfamily Associated Protein-3
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit