Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPRK1 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124535

Catalog No. PIPA124535


Add to cart

Description

Description

PA1-24535 has been successfully used in Immunohistochemistry (paraffin) procedures. PA1-24535 immunogen corresponds to Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 88/100%. Rat: 88/100%.

KOR1, an Opioid Receptor, plays a modulatory role in specific aspects of opioid function such as the control of pain. Its endogenous ligand, dynorphin A, is an important neurotransmitter in the central and peripheral nervous systems. KOR1 is believed to mediate sedation, analgesia, and diuresis. KOR1 has been reported to be expressed in the central and peripheral nervous systems, placenta, uterus, breast, and blood. ESTs have been isolated from placenta and prostate libraries.
Specifications

Specifications

OPRK1
Polyclonal
Unconjugated
Oprk1
Liquid
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18387, 29335
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
P33534, P34975
OPRK1
Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 88/100%. Rat: 88/100%.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit