Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KPNA3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528415

Catalog No. PIPA528415


Add to cart

Description

Description

PA5-28415 targets karyopherin alpha 3 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28415 immunogen is synthetic peptide corresponding to a region within amino acids 459 and 521 of Human karyopherin alpha 3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
Specifications

Specifications

KPNA3
Polyclonal
Unconjugated
KPNA3
Liquid
KPNA3
Synthetic peptide corresponding to a region within amino acids 459 and 521 of Human karyopherin alpha 3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
O00505
IPOA4, SRP1, SRP1gamma, SRP4, hSRP1, SRP1-gamma, importin alpha 4, importin alpha Q2, importin alpha-3, importin subunit alpha-3, importin-alpha-Q2, karyopherin subunit alpha-3, qip2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3839
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit