Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karyopherin β Mouse, Unlabeled, Clone: 23, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610559

 View more versions of this product

Catalog No. BDB610559


Add to cart

Description

Description

Using nuclear import assays, several cytosolic proteins that form a multi-protein complex have been identified. These proteins of 54kDa, 56kDa, and 97kDa stimulate the nuclear transport of proteins containing a nuclear localization signal (NLS). The 97kDa protein has been named karyopherin β. The NLS binds to karyopherin α. Binding is enhanced by karyopherin β. The NLS substrate also binds to the N-terminal region of karyopherin β. Both karyopherins bind to repeat sequences of nucleoporins at the nuclear envelope. Once the substrate is docked to the nuclear envelope, Ran hydrolyzes GTP and translocation occurs. Karyopherin β also binds to and inhibits the Ran GTPase protein, thus providing a mechanism of nuclear transport termination.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Karyopherin β
23
Unconjugated
Affinity Purified
Rat Karyopherin β aa. 48-241
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.