Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL1, Biotin, Polyclonal, Invitrogen™

Catalog No. PIPA132925
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA132925 50 μg
1 options

Catalog No. PIPA132925

Supplier: Thermo Scientific PA132925

Rabbit Polyclonal Antibody

PA1-32925 detects KC from mouse samples.PA1-32925 has been successfully used in Western blot applications.The PA1-32925 immunogen is: Recombinant mouse KC
TRUSTED_SUSTAINABILITY

Specifications

Antigen KC
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Gene Cxcl1
Gene Accession No. P12850
Gene Symbols CXCL1
Host Species Rabbit
Immunogen Recombinant mouse KC
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14825
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.