Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KChIP2 Rabbit anti-Rabbit, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1927

Catalog No. PA1927


Add to cart

Description

Description

PA1-927 detects Kv channel interacting protein 2 (KChIP2) b and c from rat and rabbit samples as well as recombit rat KChIP 2c protein. This antibody does not detect KChIP 2a. PA1-927 has been successfully used in Western blot procedures. By Western blot, this antibody detects ~33 and 26 kDa proteins representing KChIP2b and c, respectively, from rat ventricular myocyte whole cell lysate. The PA1-927 synthetic peptide corresponds to amino acid residues 6-24 from human KChIP2. This sequence is completely conserved in mouse and rat. The PA1-927 immunizing peptide (Cat. No. PEP-160) is available for use in neutralization and control experiments.

Voltage-gated potassium (Kv) channels are key components of the potassium currents in the heart and central nervous system. In the heart, Kv4 channels are involved in the repolarization phase of the action potential. In the brain however, these channels prevent reverse-propagation of action potentials. Found associated with Kv4 channels are a group of calcium-binding proteins termed KChIPs (Kv channel interacting proteins). KChIPs are small molecular weight proteins that bind to the cytoplasmic amino termini of Kv4 alpha-subunits and help to modulate its function. There are currently three known KChIPs; KChIP1 expressed in the brain; KChIP2 expressed in the brain, heart, and lung, and has three isoforms, a, b, and c; and KChIP3, also known as calsenilin and DREAM (downstream regulatory elements (DRE)-antagonist modulator) which is expressed in the brain and testis.
Specifications

Specifications

KChIP2
Polyclonal
Unconjugated
Kcnip2
Liquid
KCNIP2
Synthetic Peptide: R(6) K E S L S D S R D L D G S Y D Q L T(24) C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rabbit, Rat
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q9JM59
Potassium Channel Interacting Protein 2
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
100009019, 56817
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit