Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KChIP3 Rabbit anti-Rabbit, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1928

Catalog No. PA1928


Add to cart

Description

Description

PA1-928 detects Kv channel interacting protein 3 (KChIP3) from rabbit cerebellar granule cells. PA1-928 has been successfully used in immunohistochemistry procedures. Immunohistochemical staining of KChIP3 in rabbit granule cells yields a punctate staining pattern at the plasma membrane. The PA1-928 synthetic peptide corresponds to amino acid residues 30-45 from mouse KChIP3. This sequence is completely conserved in rat and 87% conserved in humans. The PA1-928 immunizing peptide (Cat. No. PEP-161) is available for use in neutralization and control experiments.

Voltage-gated potassium (Kv) channels are key components of the potassium currents in the heart and central nervous system. In the heart, Kv4 channels are involved in the repolarization phase of the action potential. In the brain however, these channels prevent reverse-propagation of action potentials. Found associated with Kv4 channels are a group of calcium-binding proteins termed KChIPs (Kv channel interacting proteins). KChIPs are small molecular weight proteins that bind to the cytoplasmic amino termini of Kv4 alpha-subunits and help to modulate its function. There are currently three known KChIPs; KChIP1 expressed in the brain; KChIP2 expressed in the brain, heart, and lung, and has three isoforms, a, b, and c; and KChIP3, also known as calsenilin and DREAM (downstream regulatory elements (DRE)-antagonist modulator) which is expressed in the brain and testis. KChIP3 has activities beyond the modulation of Kv4 currents. It has been shown that KChIP3 binds to the regulatory DRE and represses transcription from the early-response gene c-fos. Thus, KCHiP3 is the first known calcium-binding protein to function as a DNA-binding transcriptional regulator. KChIP3 is also found associated with the presenilins and is implicated in the mediation of beta-amyloid formation.
Specifications

Specifications

KChIP3
Polyclonal
Unconjugated
KCNIP3
Potassium Channel Interacting Protein 3
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
100356268
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Liquid
KCNIP3
Synthetic Peptide: I(30) K W Q R P R F T R Q A L M R C(45)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rabbit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit