Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNG3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524876

Catalog No. PIPA524876


Add to cart

Description

Description

This antibody is predicted to react with rat based on sequence homology.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member is a gamma subunit functioning as a modulatory molecule.
Specifications

Specifications

KCNG3
Polyclonal
PBS with 0.09% sodium azide
Q8TAE7, P59053
KV10.1, KV6.3, potassium voltage-gated channel subfamily G member 3, voltage-gated potassium channel 6.3, voltage-gated potassium channel Kv10.1, voltage-gated potassium channel subunit Kv10.1, voltage-gated potassium channel subunit Kv6.3, voltage-gated potassium channel su
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
170850, 225030
Immunohistochemistry, Western Blot
Unconjugated
KCNG3
Liquid
KCNG3
KLH conjugated synthetic peptide between 6-34 amino acids from the N-terminal region of human KCNG3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit