Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNH4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524371

Catalog No. PIPA524371

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. The gene is brain-specific, and located in the neocortex and the striatum. It may be involved in cellular excitability of restricted neurons in the central nervous system.
Specifications

Specifications

KCNH4
Polyclonal
PBS with 0.09% sodium azide
Q9UQ05
BEC2, ELK1, Kv12.3, ELK channel 1, brain-specific eag-like channel 2, ether-a-go-go K(+) channel family member, ether-a-go-go-like potassium channel 1, potassium voltage-gated channel subfamily H member 4, voltage-gated potassium channel subunit Kv12.3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23415
Flow Cytometry, Western Blot
Unconjugated
KCNH4
Liquid
KCNH4
KLH conjugated synthetic peptide between 67-96 amino acids from the N-terminal region of human KCNH4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit