Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNH7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526177

Catalog No. PIPA526177


Add to cart

Description

Description

KCNH7 represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This protein encodes a member of the potassium channel, voltage-gated, subfamily H.
Specifications

Specifications

KCNH7
Polyclonal
PBS with 0.09% sodium azide
Q9NS40
ERG3, HERG3, Kv11.3, ERG-3, eag related protein 3, eag-related protein 3, ether-a-go-go related gene potassium channel 3, ether-a-go-go-related gene potassium channel 3, ether-a-go-go-related protein 3, potassium channel subunit HERG-3, potassium voltage-gated channel subfamily H me
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
90134
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KCNH7
Liquid
KCNH7
KLH conjugated synthetic peptide between 52-78 amino acids from the N-terminal region of human KCNH7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit