Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KCNJ1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136965

Catalog No. NBP136965


Add to cart

Description

Description

KCNJ1 Polyclonal antibody specifically detects KCNJ1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

KCNJ1
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000
NP_000211.1
ATP-regulated potassium channel ROM-K, ATP-sensitive inward rectifier potassium channel 1, inwardly rectifying K+ channel, Kir1.1, potassium inwardly-rectifying channel, subfamily J, member 1, ROMK1inwardly rectifying subfamily J member 1
Goat
IgG
Immunogen affinity purified
RUO
Primary
3758
Human
ELISA, Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
KCNJ1
Peptide with sequence C-DQININFVVDAGNEN corresponding to internal region according to NP_722448.1.
44.8 kDa
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize reported isoforms NP_722449.2 and NP_000211. The following reported variants represent identical protein: NP_722451.1, NP_722449.2, NP_722450.1, NP_722448.1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only