Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir3.2 (KCNJ6) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525747

Catalog No. PIPA525747

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins and may be involved in the regulation of insulin secretion by glucose. It associates with two other G-protein-activated potassium channels to form a heteromultimeric pore-forming complex.
Specifications

Specifications

Kir3.2 (KCNJ6)
Polyclonal
PBS with 0.09% sodium azide
P48051
BIR1, GIRK-2, GIRK2, KATP-2, KATP2, KCNJ7, KIR3.2, hiGIRK2, G protein-activated inward rectifier potassium channel 2, inward rectifier K(+) channel Kir3.2, inward rectifier potassium channel KIR3.2, potassium channel, inwardly rectifying subfamily J member 6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3763
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
KCNJ6
Liquid
KCNJ6
KLH conjugated synthetic peptide between 26-55 amino acids from the central region of human KCNJ6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit