Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KCNK17, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192041

Catalog No. NBP192041


Add to cart

Description

Description

KCNK17 Polyclonal antibody specifically detects KCNK17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

KCNK17
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Acid-sensitive potassium channel protein TASK-4, K2p17.1, potassium channel subfamily K member 17, potassium channel, subfamily K, member 17, TALK22P domain potassium channel Talk-2, TALK-2TWIK-related alkaline pH-activated K(+) channel 2, TASK-4, TASK4TWIK-related acid-sensitive K(+) channel 4
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Affinity Purified
KCNK17
This antibody was developed against Recombinant Protein corresponding to amino acids:CSCCHHSSKEDFKSQSWRQGPDREPESHSPQQGCYPEGPMGIIQHLEPSAHAAGCGKDS
Immunogen affinity purified
RUO
Primary
89822
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only