Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNS2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA529511

Catalog No. PIPA529511


Add to cart

Description

Description

PA5-29512 targets Cytokeratin 33B in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-29512 immunogen is recombit fragment corresponding to a region within amino acids 140 and 404 of Human Cytokeratin 33B.

Potassium channel subunit. Modulates channel activity and reduces the ion flow.
Specifications

Specifications

KCNS2
Polyclonal
Unconjugated
KCNS2
Liquid
KCNS2
Recombit fragment corresponding to a region within amino acids 175 and 470 of Human KCNS2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.66 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q9ULS6
KV9.2, delayed-rectifier K(+) channel alpha subunit 2, potassium voltage-gated channel subfamily S member 2, voltage-gated potassium channel subunit Kv9.2
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3788
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit