Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD21 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525323

Catalog No. PIPA525323

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

KCTD21
Polyclonal
PBS with 0.09% sodium azide
Q4G0X4, Q3URF8
KCASH2, BTB/POZ domain-containing protein KCTD21
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
283219, 622320
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
KCTD21
Liquid
KCTD21
KLH conjugated synthetic peptide between 232-260 amino acids from the C-terminal region of human KCTD21
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit