Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Receptor 2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524710

Catalog No. PIPA524710


Add to cart

Description

Description

KDR is a major growth factor for endothelial cells. This protein encodes one of the two receptors of the KDR. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc.
Specifications

Specifications

VEGF Receptor 2
Polyclonal
Unconjugated
KDR
Liquid
KDR
KLH conjugated synthetic peptide between 1153-1182 amino acids from human KDR
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
0.48 mg/mL
PBS with 0.09% sodium azide
P35968, P35918
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2, CD309, KDR
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
16542, 3791
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit