Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KELL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528365

Catalog No. PIPA528365


Add to cart

Description

Description

PA5-28365 targets KEL in WB applications and shows reactivity with Human samples. The PA5-28365 immunogen is recombit fragment corresponding to a region within amino acids 28 and 256 of Human KEL. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases.
Specifications

Specifications

KELL
Polyclonal
Unconjugated
KEL
Liquid
KEL
Recombit fragment corresponding to a region within amino acids 28 and 256 of Human KEL
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.4 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P23276
ECE3, kell blood group antigen, kell blood group glycoprotein, KEL, Blood Group Kell Antigen
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3792
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit