Learn More
Cytokeratin 17 Mouse anti-Human, Rat, Clone: Ks 17.E3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA135545
Catalog No. 01-671-383
Add to cart
Description
Description
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
Specifications
Specifications
Cytokeratin 17
Monoclonal
Unconjugated
KRT17
Liquid
KRT17
Human cytokeratin 17.
Purified
RUO
Antibody
Monoclonal
Human, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Ks 17.E3
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
Q04695, Q6IFU8
K17, PC, PC2, PCHC1
Mouse
IgG2b
1 mL
4° C
Primary
287702, 3872