Learn More
Cytokeratin 2e Mouse anti-Bovine, Human, Mouse, Rat, Clone: Ks2.342.7.1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA135557
Catalog No. 01-671-395
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
Specifications
Specifications
Cytokeratin 2e
Monoclonal
Unconjugated
KRT2
Liquid
KRT2
Synthetic human cytokeratin 2e.
1 mL
4° C
Primary
16681, 3849, 406228, 786381
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Ks2.342.7.1
tissue culture supernatant with no preservative
P35908, Q3TTY5, Q6IG02
KRT2A, KRT2E, KRTE, MGC116967, MGC116968
Mouse
IgG1
RUO
Antibody
Monoclonal
Bovine, Human, Mouse, Rat