Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 31 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522236

Catalog No. PIPA522236


Add to cart

Description

Description

PA5-22236 targets keratin 31 in WB applications and shows reactivity with Human samples. The PA5-22236 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 261 of Cytokeratin 31 (Uniprot IDNo.Q15323). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Cytokeratin 31
Polyclonal
Unconjugated
KRT31
Liquid
KRT31
Recombit fragment corresponding to a region within amino acids 1 and 261 of Cytokeratin 31 (Uniprot IDNo.Q15323)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.3 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q15323
HA1, Ha-1, KRTHA1, hHa1, K31, hair keratin, type I Ha1, hard keratin, type I, 1, keratin, hair, acidic,1, keratin, type I cuticular Ha1, keratin-31
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3881
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit