Learn More
Cytokeratin 8 Mouse anti-Human, Clone: M20, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA135565
Catalog No. 01-671-003
Add to cart
Description
Description
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
Specifications
Specifications
Cytokeratin 8
Monoclonal
Unconjugated
KRT8
Liquid
KRT8
Cytokeratin purified from human breast carcinoma cell line MC-7.
1 mL
4° C
Primary
3856
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
M20
tissue culture supernatant with 1% FBS and 10mM sodium azide
P05787
CARD2, CK8, CYK8, K2C8, K8, KO
Mouse
IgG1
RUO
Antibody
Monoclonal
Human